Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PPP2R5B Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

Supplier:  Novus Biologicals NBP18895825UL

 View more versions of this product

Catalog No. NB431006


Only null left
Add to Cart

Description

Description

PPP2R5B Polyclonal specifically detects PPP2R5B in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

PPP2R5B
Polyclonal
Western Blot 0.4 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin 1:500 - 1:1000
B56B, FLJ35411, PP2A B subunit isoform B56-beta, PP2A B subunit isoform B'-beta, PP2A B subunit isoform PR61-beta, PP2A B subunit isoform R5-beta, PR61B, protein phosphatase 2, regulatory subunit B (B56), beta isoform, protein phosphatase 2, regulatory subunit B', beta, protein phosphatase 2, regulatory subunit B', beta isoform, serine/threonine protein phosphatase 2A, 56 kDa regulatory subunit, betaisoform, serine/threonine-protein phosphatase 2A 56 kDa regulatory subunit beta isoform
Rabbit
Affinity Purified
RUO
Primary
Specificity of human PPP2R5B antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Unconjugated
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
PPP2R5B
This antibody was developed against Recombinant Protein corresponding to amino acids:GKLFDELTASYKLEKQQEQQKAQERQELWQGLEELRLRRLQGTQGAKEAPLQRLTP
25 μL
Wnt Signaling Pathway
5526
Human, Mouse, Rat
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.

For Research Use Only