Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PPP2R5B Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $646.00
Specifications
Antigen | PPP2R5B |
---|---|
Dilution | Western Blot 0.4 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin 1:500 - 1:1000 |
Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
PPP2R5B Polyclonal specifically detects PPP2R5B in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
PPP2R5B | |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
Human, Mouse, Rat | |
B56B, FLJ35411, PP2A B subunit isoform B56-beta, PP2A B subunit isoform B'-beta, PP2A B subunit isoform PR61-beta, PP2A B subunit isoform R5-beta, PR61B, protein phosphatase 2, regulatory subunit B (B56), beta isoform, protein phosphatase 2, regulatory subunit B', beta, protein phosphatase 2, regulatory subunit B', beta isoform, serine/threonine protein phosphatase 2A, 56 kDa regulatory subunit, betaisoform, serine/threonine-protein phosphatase 2A 56 kDa regulatory subunit beta isoform | |
PPP2R5B | |
IgG | |
Affinity Purified | |
Specificity of human PPP2R5B antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Western Blot 0.4 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin 1:500 - 1:1000 | |
Polyclonal | |
Rabbit | |
Wnt Signaling Pathway | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
5526 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:GKLFDELTASYKLEKQQEQQKAQERQELWQGLEELRLRRLQGTQGAKEAPLQRLTP | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title