Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PPP2R5D Antibody, Novus Biologicals™
SDP

Catalog No. NB100846 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB100846 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB100846 Supplier Novus Biologicals Supplier No. NB100846
Add to Cart
Add to Cart

Goat Polyclonal Antibody

PPP2R5D Polyclonal specifically detects PPP2R5D in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, Peptide ELISA, Western Blot.

Specifications

Antigen PPP2R5D
Applications Flow Cytometry, Immunocytochemistry, Immunofluorescence, Peptide ELISA, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.03 - 0.3 ug/mL, Flow Cytometry 10 ug/mL, Immunocytochemistry/Immunofluorescence 10 ug/mL, Peptide ELISA Detection limit 1:20000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. NP_006236.
Gene Alias MGC2134, MGC8949, PP2A B subunit isoform B56-delta, PP2A B subunit isoform B'-delta, PP2A B subunit isoform PR61-delta, PP2A B subunit isoform R5-delta, PP2A, B subunit, B' delta isoform, PP2A, B subunit, B56 delta isoform, PP2A, B subunit, PR61 delta isoform, PP2A, B subunit, R5 delta isoform, protein phosphatase 2, regulatory subunit B', delta, regulatory subunit B (B56), delta isoform, regulatory subunit B', delta isoform, Serine/threonine protein phosphatase 2A, 56 kDa regulatory subunit, deltaisoform, serine/threonine-protein phosphatase 2A 56 kDa regulatory subunit delta isoform
Gene Symbols PPP2R5D
Host Species Goat
Immunogen Peptide with sequence C-KRAEEFLTASQEAL corresponding to C-Terminus according to NP_006236.1, NP_851307.1, NP_851308.1.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Neuroscience, Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 5528
Test Specificity This antibody is expected to recognize all three reported isoforms (NP_006236.1; NP_851307.1; NP_851308.1).
Target Species Human, Mouse, Rat
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.