Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PPP2R5D Antibody (OTI5E7), Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB301845 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB301845 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB301845 Supplier Novus Biologicals Supplier No. NBP273587APC
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PPP2R5D Monoclonal antibody specifically detects PPP2R5D in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry

Specifications

Antigen PPP2R5D
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI5E7
Conjugate APC
Dilution Western Blot, Immunohistochemistry
Formulation PBS
Gene Alias MGC2134, MGC8949, PP2A B subunit isoform B56-delta, PP2A B subunit isoform B'-delta, PP2A B subunit isoform PR61-delta, PP2A B subunit isoform R5-delta, PP2A, B subunit, B' delta isoform, PP2A, B subunit, B56 delta isoform, PP2A, B subunit, PR61 delta isoform, PP2A, B subunit, R5 delta isoform, protein phosphatase 2, regulatory subunit B', delta, regulatory subunit B (B56), delta isoform, regulatory subunit B', delta isoform, Serine/threonine protein phosphatase 2A, 56 kDa regulatory subunit, deltaisoform, serine/threonine-protein phosphatase 2A 56 kDa regulatory subunit delta isoform
Host Species Mouse
Immunogen Full length human recombinant protein of human PPP2R5D (NP_006236) produced in E.coli.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience, Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 5528
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.