Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PPP3CC Antibody, Novus Biologicals™
SDP

Catalog No. NBP219911 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP219911 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP219911 Supplier Novus Biologicals Supplier No. NBP219911
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PPP3CC Polyclonal specifically detects PPP3CC in Human samples. It is validated for Western Blot.

Specifications

Antigen PPP3CC
Applications Western Blot
Classification Polyclonal
Concentration 0.55mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000-1:10000
Formulation 0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal
Gene Accession No. P48454
Gene Alias Calcineurin, testis-specific catalytic subunit, Calmodulin-dependent calcineurin A subunit gamma isoform, CALNA3CAM-PRP catalytic subunit, CNA3, EC 3.1.3.16, PP2Bgamma, protein phosphatase 2B, catalytic subunit, gamma isoform, protein phosphatase 3 (formerly 2B), catalytic subunit, gamma isoform, protein phosphatase 3 (formerly 2B), catalytic subunit, gamma isoform(calcineurin A gamma), protein phosphatase 3, catalytic subunit, gamma isozyme, serine/threonine-protein phosphatase 2B catalytic subunit gamma isoform
Gene Symbols PPP3CC
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human PPP3CC. The exact sequence is proprietary.
Molecular Weight of Antigen 58 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5533
Test Specificity Human. Species reactivity based on sequence identity: Mouse (93%), Rabbit (84%), Rat (93%), Bovine (97%), Xenopus tropicalis (86%).
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.