Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PPP4R4 Antibody, Novus Biologicals™
SDP

Catalog No. NBP219917 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP219917 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP219917 Supplier Novus Biologicals Supplier No. NBP219917
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PPP4R4 Polyclonal specifically detects PPP4R4 in Human samples. It is validated for Western Blot.

Specifications

Antigen PPP4R4
Applications Western Blot
Classification Polyclonal
Concentration 1mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000
Formulation PBS (pH 7.0) and 20% Glycerol with 0.01% Thimerosal
Gene Accession No. Q6NUP7
Gene Alias KIAA1622PP4R4HEAT-like repeat-containing protein, protein phosphatase 4, regulatory subunit 4, serine/threonine-protein phosphatase 4 regulatory subunit 4
Gene Symbols PPP4R4
Host Species Rabbit
Immunogen Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human PP4R4. The exact sequence is proprietary.
Molecular Weight of Antigen 99 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57718
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.