Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PPP6C Antibody (SR1537), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP322483100UL
Description
PPP6C Monoclonal antibody specifically detects PPP6C in Human samples. It is validated for Western Blot, Flow Cytometry, ImmunofluorescenceSpecifications
PPP6C | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Flow Cytometry, Immunofluorescence | |
SR1537 | |
Western Blot 1:500-1:2000, Flow Cytometry 1:50, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
EC 3.1.3.16, FLJ92648, MGC12249, PP6, PP6C, PPP6, protein phosphatase 6, catalytic subunit, serine/threonine protein phosphatase catalytic subunit, serine/threonine-protein phosphatase 6 catalytic subunit | |
A synthesized peptide derived from human PPP6C (Uniprot #: O00743) | |
100 μg | |
Cell Cycle and Replication, Signal Transduction | |
5537 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction