Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PPP6C Antibody (SR1537), Novus Biologicals™

Rabbit Monoclonal Antibody
$309.50 - $423.00
Specifications
Antigen | PPP6C |
---|---|
Clone | SR1537 |
Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:50, Immunocytochemistry/Immunofluorescence 1:50-1:200 |
Applications | Western Blot, Flow Cytometry, Immunofluorescence |
Classification | Monoclonal |
Description
PPP6C Monoclonal antibody specifically detects PPP6C in Human samples. It is validated for Western Blot, Flow Cytometry, ImmunofluorescenceSpecifications
PPP6C | |
Western Blot 1:500-1:2000, Flow Cytometry 1:50, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
EC 3.1.3.16, FLJ92648, MGC12249, PP6, PP6C, PPP6, protein phosphatase 6, catalytic subunit, serine/threonine protein phosphatase catalytic subunit, serine/threonine-protein phosphatase 6 catalytic subunit | |
A synthesized peptide derived from human PPP6C (Uniprot #: O00743) | |
Primary | |
Store at -20° C. Avoid freeze/thaw cycles. |
SR1537 | |
Western Blot, Flow Cytometry, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Cell Cycle and Replication, Signal Transduction | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
5537 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title