Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PPWD1 Antibody (OTI1B8), Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NP27390A30 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP27390A30 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP27390A30 Supplier Novus Biologicals Supplier No. NBP273590AF350
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PPWD1 Monoclonal specifically detects PPWD1 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen PPWD1
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone OTI1B8
Conjugate Alexa Fluor 350
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias EC 5.2.1.8, KIAA0073Spliceosome-associated cyclophilin, peptidylprolyl isomerase domain and WD repeat containing 1, peptidylprolyl isomerase domain and WD repeat-containing protein 1
Gene Symbols PPWD1
Host Species Mouse
Immunogen Full length recombinant protein of human PPWD1 (NP_056157) produced in HEK293T cells.
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 23398
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.