Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ PPY A

Click to view available options
Quantity:
10 mg
50 mg
Description
Potent inhibitor of T315l mutant and wild-type Abl kinases (IC50 values are 9 and 20 nM, respectively). Inhibits growth of cells transformed with either the Bcr-Abl T315l mutant or wild-type Bcr-Abl gene.
Specifications
Specifications
| Quantity | 10 mg |
| Formula Weight | 372.42 |
| Purity | >98% |
| Molecular Weight (g/mol) | 372.428 |
| Chemical Name or Material | PPY A |
Safety and Handling
| Recommended Storage | Store at -20°C |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction