Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PQBP1 Rabbit anti-Mouse, Rat, Clone: 8D8H6, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $458.50
Specifications
Antigen | PQBP1 |
---|---|
Clone | 8D8H6 |
Applications | Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
PQBP1 Monoclonal antibody specifically detects PQBP1 in Mouse, Rat samples. It is validated for Western BlotSpecifications
PQBP1 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Cancer, Endocrinology, Neurodegeneration, Neuroscience, Signal Transduction | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
10084 | |
IgG | |
Affinity purified |
8D8H6 | |
Monoclonal | |
Purified | |
RUO | |
Mouse, Rat | |
38 kDa nuclear protein containing a WW domain, MRX55, MRXS3, MRXS8, Npw38, NPW38mental retardation, X-linked 55, nuclear protein containing WW domain 38 kD, polyglutamine binding protein 1, Polyglutamine tract-binding protein 1, polyglutamine-binding protein 1, PQBP-1, RENS1, SHS, Sutherland-Haan X-linked mental retardation syndrome | |
Recombinant protein of human PQBP1. | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title