Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PR48 Antibody, Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB292033 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB292033 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB292033 Supplier Novus Biologicals Supplier No. NB10041411APC
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PR48 Polyclonal antibody specifically detects PR48 in Human samples. It is validated for Western Blot, Immunoprecipitation

Specifications

Antigen PR48
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Conjugate APC
Dilution Western Blot, Immunoprecipitation
Formulation PBS
Gene Alias FLJ60425, NYREN8, NY-REN-8 antigen, PP2A subunit B isoform PR48, PP2A, subunit B, PR48 isoform, PPP2R3L, PPP2R3LY, PR48, protein phosphatase 2 (formerly 2A), regulatory subunit B'', beta, protein phosphatase 2, regulatory subunit B'', beta, Protein phosphatase 2A 48 kDa regulatory subunit, serine/threonine protein phosphatase 2A, 48kDa regulatory subunit B, serine/threonine-protein phosphatase 2A regulatory subunit B'' subunit beta
Host Species Rabbit
Immunogen Maps to a region between residue 350 and 400 of human protein phosphatase 2 (formerly 2A), regulatory subunit B'', beta using the numbering given in entry NP_037371.2 (GeneID 28227).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 28227
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.