Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PR48 Rabbit anti-Human, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB159860 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB159860 100 μg
NB159861 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB159860 Supplier Novus Biologicals Supplier No. NBP317915100UL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PR48 Polyclonal antibody specifically detects PR48 in Human samples. It is validated for Western Blot, Immunofluorescence

Specifications

Antigen PR48
Applications Western Blot, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 0.04-0.4 ug/ml, Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml
Formulation PBS, pH 7.2, 40% glycerol
Gene Alias FLJ60425, NYREN8, NY-REN-8 antigen, PP2A subunit B isoform PR48, PP2A, subunit B, PR48 isoform, PPP2R3L, PPP2R3LY, PR48, protein phosphatase 2 (formerly 2A), regulatory subunit B'', beta, protein phosphatase 2, regulatory subunit B'', beta, Protein phosphatase 2A 48 kDa regulatory subunit, serine/threonine protein phosphatase 2A, 48kDa regulatory subunit B, serine/threonine-protein phosphatase 2A regulatory subunit B'' subunit beta
Host Species Rabbit
Immunogen This antibody was developed against Recombinant Protein corresponding to amino acids: KTPTSIEYWFRCMDLDGDGALSMFELEYFYEEQCRRLDSMAIEALPFQDCLCQMLDLVKPRTEGKITLQDLKRCKLANVFFDTFFNIEKYLDHEQKEQISLLRDGDSGGPELSDWEKYAAEEYDILVAEETAGEPWEDGFEAELSPVEQ
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 28227
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.