Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PRAC Antibody, PE, Novus Biologicals™
SDP

Catalog No. NB332412 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB332412 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB332412 Supplier Novus Biologicals Supplier No. NBP297053PE
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PRAC Polyclonal antibody specifically detects PRAC in Human samples. It is validated for ELISA

Specifications

Antigen PRAC
Applications ELISA
Classification Polyclonal
Conjugate PE
Dilution ELISA
Formulation PBS
Gene Alias C17orf92, chromosome 17 open reading frame 92, MGC32520, PRAC1, prostate cancer susceptibility candidate, Prostate, rectum and colon expressed gene protein, small nuclear protein PRAC
Host Species Rabbit
Immunogen Rabbits immunized with purified, recombinant Human PRAC (Accession#: NP_115767.1, Met1-Phe57)
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Biology, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 84366
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.