Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PRAGMIN Antibody, Novus Biologicals™
SDP

Catalog No. NBP142689 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP142689 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP142689 Supplier Novus Biologicals Supplier No. NBP142689
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PRAGMIN Polyclonal specifically detects PRAGMIN in Human samples. It is validated for Western Blot, Immunoprecipitation.

Specifications

Antigen PRAGMIN
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:2000-1:10000, Immunoprecipitation 2-5 ug/mg lysate
Formulation TBS and 0.1% BSA with 0.09% Sodium Azide
Gene Alias DKFZp761P0423, homolog of rat pragma of Rnd2, PRAGMIN, sugen kinase 223, tyrosine-protein kinase SgK223
Gene Symbols SGK223
Host Species Rabbit
Immunogen The immunogen for this product maps to a region between residue 275 and 325 of human homolog of rat pragma of Rnd2 using the numbering given in entry NP_001074295.1 (GeneID 157285).
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 157285
Test Specificity Human.Based on 100% sequence identity, this antibody is predicted to react with Chimpanzee and Northern white-cheeked gibbon.
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.