Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PRAK Antibody (OTI5E1), FITC, Novus Biologicals™
SDP

Catalog No. NB301838 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB301838 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB301838 Supplier Novus Biologicals Supplier No. NBP273594F
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PRAK Monoclonal antibody specifically detects PRAK in Human, Mouse, Rat samples. It is validated for Western Blot

Specifications

Antigen PRAK
Applications Western Blot
Classification Monoclonal
Clone OTI5E1
Conjugate FITC
Dilution Western Blot
Formulation PBS
Gene Alias EC 2.7.11, MAP kinase-activated protein kinase 5, MAPK-activated protein kinase 5, MAPKAP kinase 5, MAPKAPK-5, mitogen-activated protein kinase-activated protein kinase 5, p38-regulated/activated protein kinase, PRAKEC 2.7.11.1
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 197-471 of human MAPKAPK5 (NP_003659) produced in E.coli.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 8550
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.