Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PRAM1 Antibody, Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NB051581 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB051581 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB051581 Supplier Novus Biologicals Supplier No. NBP298045AF350
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PRAM1 Polyclonal specifically detects PRAM1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen PRAM1
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 350
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias MGC39864, PML-RAR, PML-RARA regulated adaptor molecule 1, PML-RARA target gene encoding an Adaptor Molecule-1, PML-RARA-regulated adapter molecule 1, PRAM, PRAM-1
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human PRAM1 fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84106
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.