Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PRAME Antibody, Novus Biologicals™
SDP

Catalog No. NBP223670 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP223670 0.1 mL
NBP223670SS 0.025 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP223670 Supplier Novus Biologicals Supplier No. NBP223670
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PRAME Polyclonal specifically detects PRAME in Human samples. It is validated for Western Blot, Simple Western.

Specifications

Antigen PRAME
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1: 1000-1:5000, Simple Western 1:5000
Formulation 0.1 ml crude antibody whole antisera with 50% Glycerol with 0.025% Sodium Azide
Gene Accession No. P78395
Gene Alias cancer/testis antigen 130, MAPEOIP4CT130, melanoma antigen preferentially expressed in tumors, OIP-4, Opa-interacting protein 4, Opa-interacting protein OIP4, preferentially expressed antigen in melanoma, Preferentially expressed antigen of melanoma
Gene Symbols PRAME
Host Species Rabbit
Immunogen A portion of amino acid 460-509 of human PRAME was used as the immunogen for the antibody.
Purification Method Unpurified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 23532
Test Specificity The amino acid sequence used as immunogen for the antibody is 100% homologous in human, monkey, dog, and horse. It is 92% homologous in chimpanzee.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Antisera
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.