Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PRAME Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.50 - $426.00
Specifications
Antigen | PRAME |
---|---|
Dilution | Western Blot 1: 1000-1:5000, Simple Western 1:5000 |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP223670SS
![]() |
Novus Biologicals
NBP223670SS |
0.025 mL |
Each for $206.50
|
|
|||||
NBP223670
![]() |
Novus Biologicals
NBP223670 |
0.1 mL |
Each for $426.00
|
|
|||||
Description
PRAME Polyclonal specifically detects PRAME in Human samples. It is validated for Western Blot, Simple Western.Specifications
PRAME | |
Western Blot | |
Unconjugated | |
Rabbit | |
Cancer | |
0.1 ml crude antibody whole antisera with 50% Glycerol with 0.025% Sodium Azide | |
cancer/testis antigen 130, MAPEOIP4CT130, melanoma antigen preferentially expressed in tumors, OIP-4, Opa-interacting protein 4, Opa-interacting protein OIP4, preferentially expressed antigen in melanoma, Preferentially expressed antigen of melanoma | |
PRAME | |
IgG | |
Unpurified | |
The amino acid sequence used as immunogen for the antibody is 100% homologous in human, monkey, dog, and horse. It is 92% homologous in chimpanzee. |
Western Blot 1: 1000-1:5000, Simple Western 1:5000 | |
Polyclonal | |
Antisera | |
RUO | |
Human | |
P78395 | |
23532 | |
A portion of amino acid 460-509 of human PRAME was used as the immunogen for the antibody. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title