Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Praseodymium(III, IV) oxide, 99.5% (REO)

Catalog No. AA1123414
Click to view available options
Quantity:
25 g
100 g

Catalog No. AA1123414

Supplier: Thermo Scientific Chemicals 01123414

Only null left
Add to Cart

CAS: 12037-29-5 | O11Pr6 | 1021.44 g/mol

Praseodymium(III, IV) oxide is used in pigment in glass, optic and ceramic industries. Praseodymium-doped glass, called didymium glass is used in welding, blacksmithing, and glass-blowing goggles due to its blocking property of infrared radiation. It is employed in the solid state synthesis of praseodymium molybdenum oxide, which is used as a semiconductor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12037-29-5
Molecular Formula O11Pr6
Molecular Weight (g/mol) 1021.44
MDL Number MFCD00011178
InChI Key HPZIIFVSYNLWNX-UHFFFAOYSA-N
IUPAC Name hexapraseodymium(3+) undecaoxidandiide
SMILES [O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[Pr+3].[Pr+3].[Pr+3].[Pr+3].[Pr+3].[Pr+3]

Specifications

Melting Point 2500°C
Physical Form Powder
Quantity 25 g
Solubility Information Insoluble in water. Moderately soluble in strong mineral acids.
Formula Weight 1021.43
Percent Purity 99.5%
Assay (REO)
Chemical Name or Material Praseodymium(III, IV) oxide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H319-H335
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 234-857-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.