Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PRAT4A/TNRC5 Antibody, Novus Biologicals™
SDP

Catalog No. NBP223847 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP223847 0.1 mL
NBP223847SS 0.025 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP223847 Supplier Novus Biologicals Supplier No. NBP223847
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 1 publication

PRAT4A/TNRC5 Polyclonal specifically detects PRAT4A/TNRC5 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin).

Specifications

Antigen PRAT4A/TNRC5
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunohistochemistry-Paraffin 1: 1000-1:2000
Formulation 0.1 ml crude whole Rabbit sera with 50% Glycerol with 0.025% Sodium Azide
Gene Accession No. Q9BT09
Gene Alias CAG4A, canopy 3 homolog (zebrafish), CTG4A, ERDA5CTG repeat protein 4a, Expanded repeat-domain protein CAG/CTG 5, PRAT4ACAG repeat containing, Protein associated with TLR4, protein associated with Toll-like receptor 4A, protein canopy homolog 3, TNRC5expanded repeat domain, CAG/CTG 5, trinucleotide repeat containing 5, Trinucleotide repeat-containing gene 5 protein
Gene Symbols CNPY3
Host Species Rabbit
Immunogen A portion of amino acids 200-250 of human PRAT4A was used as the immunogen for this antibody.
Purification Method Unpurified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10695
Test Specificity The amino acid sequence used as immunogen is 100% homologous in mouse and rat.
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Antisera
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.