Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PRAT4A/TNRC5 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 1 publication
$181.90 - $402.00
Specifications
Antigen | PRAT4A/TNRC5 |
---|---|
Dilution | Immunohistochemistry-Paraffin 1: 1000-1:2000 |
Applications | Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP223847SS
![]() |
Novus Biologicals
NBP223847SS |
0.025 mL |
Each for $181.90
|
|
|||||
NBP223847
![]() |
Novus Biologicals
NBP223847 |
0.1 mL |
Each for $402.00
|
|
|||||
Description
PRAT4A/TNRC5 Polyclonal specifically detects PRAT4A/TNRC5 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
PRAT4A/TNRC5 | |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Human | |
Q9BT09 | |
10695 | |
A portion of amino acids 200-250 of human PRAT4A was used as the immunogen for this antibody. | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
Immunohistochemistry-Paraffin 1: 1000-1:2000 | |
Polyclonal | |
Antisera | |
RUO | |
0.1 ml crude whole Rabbit sera with 50% Glycerol with 0.025% Sodium Azide | |
CAG4A, canopy 3 homolog (zebrafish), CTG4A, ERDA5CTG repeat protein 4a, Expanded repeat-domain protein CAG/CTG 5, PRAT4ACAG repeat containing, Protein associated with TLR4, protein associated with Toll-like receptor 4A, protein canopy homolog 3, TNRC5expanded repeat domain, CAG/CTG 5, trinucleotide repeat containing 5, Trinucleotide repeat-containing gene 5 protein | |
CNPY3 | |
IgG | |
Unpurified | |
The amino acid sequence used as immunogen is 100% homologous in mouse and rat. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title