Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Praziquantel, USP, 98.5-101%, Spectrum™ Chemical

Click to view available options
Quantity:
25 g
Packaging:
Amber Glass Bottle
Description
Spectrum™ Chemical Praziquantel, USP belongs to a class of drugs known as antihelmintics and is used to treat infections of certain parasites. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications
Specifications
CAS Max % | 1 |
Ignition Residue | 0.001 |
Loss on Drying | 0.005 |
Packaging | Amber Glass Bottle |
Quantity | 25 g |
Percent Purity | 98.5 to 101% |
Grade | USP |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction