Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Praziquantel, USP, 98.5-101%, Spectrum™ Chemical
SDP

Catalog No. 18605779
Click to view available options
Quantity:
100 g
500 g
1 kg
25 kg
Packaging:
Amber Glass Bottle
Fiber Drum

P1641, 55268-74-1, C19H24N2O2

Spectrum™ Chemical Praziquantel, USP belongs to a class of drugs known as antihelmintics and is used to treat infections of certain parasites. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 55268-74-1
Molecular Formula C19H24N2O2
Molecular Weight (g/mol) 312.41
InChI Key FSVJFNAIGNNGKK-UHFFFAOYNA-N
IUPAC Name 2-cyclohexanecarbonyl-1H,2H,3H,4H,6H,7H,11bH-pyrazino[2,1-a]isoquinolin-4-one
SMILES O=C(C1CCCCC1)N1CC2N(CCC3=CC=CC=C23)C(=O)C1

Specifications

CAS Max % 1
Ignition Residue 0.001
Loss on Drying 0.005
Packaging Amber Glass Bottle
Quantity 100 g
Percent Purity 98.5 to 101%
Grade USP
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.