Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PRB3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | PRB3 |
---|---|
Concentration | 0.4 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-3762
![]() ![]() |
Proteintech Group Inc
221901AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
PRB3 protein is identical to the gross cystic disease fluid protein GCDFP-15, secretory actin binding protein (SABP) and glycoprotein 17 (GP-17). GCDFP-15 is expressed by breast carcinomas (primary and metastatic lesions) with apocrine features.Specifications
PRB3 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
Q04118 | |
5544, 639802 | |
PRB3 Fusion Protein Ag16760 | |
Primary | |
-20°C | |
PRB3 |
0.4 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
G1, PRB3, PRG, Proline rich protein G1 | |
PRB3, Prb3-ps | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title