Learn More
PRCC Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc IHC00675

Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Orangutan, Monkey, Gorilla, Chimpanzee and Northern white-cheeked gibbon
PRCC may regulate cell cycle progression through interaction with MAD2L2.Specifications
PRCC | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q92733 | |
PRCC | |
Between 125 and 175 | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Immunohistochemistry | |
Unconjugated | |
PRCC | |
papillary renal cell carcinoma translocation-associated gene protein, RCCP1, TPRC | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
5546, 94315 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.