Learn More
PRCC Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
$151.31 - $568.26
Specifications
Antigen | PRCC |
---|---|
Applications | Immunohistochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Liquid |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-157-2518
![]() |
Bethyl Laboratories, Inc
IHC00675T |
10 μL |
Each for $151.31
|
|
|||||
50-157-2517
![]() |
Bethyl Laboratories, Inc
IHC00675 |
100 μL |
Each for $568.26
|
|
|||||
Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Orangutan, Monkey, Gorilla, Chimpanzee and Northern white-cheeked gibbon
PRCC may regulate cell cycle progression through interaction with MAD2L2.Specifications
PRCC | |
Polyclonal | |
Liquid | |
RUO | |
TBS with 0.1% BSA and 0.09% sodium azide | |
papillary renal cell carcinoma translocation-associated gene protein, RCCP1, TPRC | |
Between 125 and 175 | |
Primary | |
4° C | |
PRCC |
Immunohistochemistry | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
Q92733 | |
5546, 94315 | |
IgG | |
Antigen affinity chromatography | |
Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.