Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PRCP Polyclonal Antibody
Rabbit Polyclonal Antibody
$209.50 - $511.50
Specifications
Antigen | PRCP |
---|---|
Concentration | 0.5 mg/mL |
Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIPA5119360
|
Invitrogen™
PA5119360 |
200 μL |
Each for $511.50
|
|
|||||
PIPA5121671
|
Invitrogen™
PA5121671 |
20 μL |
Each for $209.50
|
|
|||||
Description
PRCP Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
Cleaves C-terminal amino acids linked to proline in peptides such as angiotensin II, III and des-Arg9-bradykinin. This cleavage occurs at acidic pH, but enzymatic activity is retained with some substrates at neutral pH.Specifications
PRCP | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Polyclonal | |
Liquid | |
IgG | |
Mouse | |
PRCP | |
2510048K03Rik; 2610104A14Rik; AI451719; Angiotensinase C; HUMPCP; Lysosomal carboxypeptidase C; lysosomal Pro-X carboxypeptidase; PCP; Prcp; Proline carboxypeptidase; prolylcarboxypeptidase; prolylcarboxypeptidase (angiotensinase C) | |
PRCP | |
Primary | |
Antigen affinity chromatography, Protein A |
0.5 mg/mL | |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
RUO | |
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
Q7TMR0 | |
72461 | |
Recombinant protein Prolylcarboxypeptidase. The antigen corresponds to amino acid range 169-443 of the target protein. | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title