Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PRDM5 Antibody, Novus Biologicals™
SDP

Catalog No. NB10056419T Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10056419T 0.025 mg
NB10056419 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB10056419T Supplier Novus Biologicals Supplier No. NB10056419SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PRDM5 Polyclonal specifically detects PRDM5 in Human samples. It is validated for Western Blot.

Specifications

Antigen PRDM5
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1-3 ug/ml
Formulation PBS containing 0.2% gelatin with 0.05% Sodium Azide
Gene Accession No. Q9NQX1
Gene Alias PFM2PR domain-containing protein 5, PR domain containing 5, PR domain zinc finger protein 5
Gene Symbols PRDM5
Host Species Rabbit
Immunogen A portion of amino acids 100-200 of human PRDM5 protein were used as the immunogen.
Purification Method Protein G purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Chromatin Research, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 11107
Test Specificity Cross reacts with Human. Predicted to cross react with Canine.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.