Learn More
Description
Includes
20 μl for 20 gels
- The sizes of the λ / EcoRI DNA fragments, in base pairs, are 21226, 7421, 5804, 5643, 4878, and 3530
- A sample volume of 20 μl is recommended for ethidium bromide or SYBR Safe staining and 35-40 μl is optimal for methylene blue staining
- Shortly before loading, heat the fragments at 65°C for 3 minutes and quickly chill on ice
Specifications
Specifications
| Device Holds | 20 Gels |
| Type | Lambda DNA Digested |
| Includes | Eco RI |
| Quantity | 20 g |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

