Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pref-1/DLK1/FA1 Antibody (PF13-3), Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. NB172451 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB172451 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB172451 Supplier Novus Biologicals Supplier No. NBP280046AF405
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Pref-1/DLK1/FA1 Monoclonal antibody specifically detects Pref-1/DLK1/FA1 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)

Specifications

Antigen Pref-1/DLK1/FA1
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone PF13-3
Conjugate Alexa Fluor 405
Formulation 50mM Sodium Borate
Gene Alias delta-like 1 homolog (Drosophila), DLK, DLK-1, FA1fetal antigen 1, pG2delta-like homolog (Drosophila), preadipocyte factor 1, Pref-1, PREF1, protein delta homolog 1, secredeltin, ZOG
Host Species Mouse
Immunogen Recombinant human Pref-1/DLK1/FA1 (extracellular domain).
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 8788
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.