Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pref-1/DLK1/FA1, Mouse anti-Human, Clone: PF13-3, Novus Biologicals™
SDP

Catalog No. NB013169 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB013169 50 μg
NB013168 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB013169 Supplier Novus Biologicals Supplier No. NBP28004650UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Pref-1/DLK1/FA1 Monoclonal specifically detects Pref-1/DLK1/FA1 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, CyTOF-ready.

Specifications

Antigen Pref-1/DLK1/FA1
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry
Classification Monoclonal
Clone PF13-3
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot, Flow Cytometry, ELISA, Immunohistochemistry
Formulation 0.2 um-filtered solution in PBS, pH 7.4
Gene Alias delta-like 1 homolog (Drosophila), DLK, DLK-1, FA1fetal antigen 1, pG2delta-like homolog (Drosophila), preadipocyte factor 1, Pref-1, PREF1, protein delta homolog 1, secredeltin, ZOG
Gene Symbols DLK1
Host Species Mouse
Immunogen Recombinant human Pref-1/DLK1/FA1(extracellular domain).
Purification Method Protein G purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8788
Test Specificity Recognizes human Pref-1/DLK1/FA1. Does not cross-react with mouse Pref-1/DLK1/FA1.
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.