Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abnova™ PRELP 293T Cell Transient Overexpression Lysate (Denatured) (T01)
Click to view available options
Quantity:
100 μL
Specifications
Specifications
Cell Line | 293T |
For Use With (Application) | Western Blot |
Description | PRELP 293T Cell Transient Overexpression Lysate(Denatured) |
Gene Accession No. | NM_002725 |
Molecular Weight (g/mol) | Theoretical M.W. 43.8kDa |
Plasmid | pCMV-PRELP full-length |
Quality Control Testing | Transient overexpression cell lysate was tested with anti-PRELP antibody (Mfr. No. H00005549-B01) by Western Blot. |
Quantity | 100 μL |
Storage Buffer | 1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue) |
Host Species | Human |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction