Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PRELP Antibody, Janelia Fluor™ 549, Novus Biologicals™
SDP

Catalog No. NB322332 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB322332 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB322332 Supplier Novus Biologicals Supplier No. NBP300141JF549
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PRELP Polyclonal antibody specifically detects PRELP in Human samples. It is validated for ELISA

Specifications

Antigen PRELP
Applications ELISA
Classification Polyclonal
Conjugate Janelia Fluor 549
Dilution ELISA
Formulation 50mM Sodium Borate
Gene Alias 55 kDa leucine-rich repeat protein of articular cartilage, MST161, MSTP161, prolargin, prolargin proteoglycan, proline arginine-rich end leucine-rich repeat protein, proline/arginine-rich end leucine-rich repeat protein, Proline-arginine-rich end leucine-rich repeat protein, SLRR2AMGC45323
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Human PRELP (Accession#: NP_002716.1; Gln21-Ile382)
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5549
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.