Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Molecular Probes™ Premo™ Halide Sensor

Catalog No. P10229
Encompass_Preferred
Change view
Click to view available options
Quantity:
20 mL Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P10229 20 mL Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P10229 Supplier Molecular Probes™ Supplier No. P10229
Add to Cart
Edge
Add to Cart

Premo™ Halide Sensor

The Premo™ Halide Sensor is based on a yellow fluorescent protein (YFP) molecule sensitive to halide ions. The combination of the YFP (Venus) biosensor with efficient and non-cytopathic BacMam delivery and expression gives researchers a highly sensitive, robust and easy-to-use tool to efficiently screen halide ion channels and transporter modulators in their cellular models of choice. No specialized instruments are needed, as the sensor works on standard HTS platforms. - Cl– channel–specific—measure halide ion flux in voltage- and ligand-gated chloride channels and transporters - Fast—measure chloride flux in high-throughput mode with highly reproducible results - Robust—reliably high expression of the bright and fast maturating Venus sensor gives an excellent signal window - Convenient—study chloride flux in your cellular model by efficient and non-cytopathic delivery of Premo™ Halide Sensor by BacMam technology - Pharmacologically relevant—known modulators show dose-dependent quenching and BacMam delivery enables assays in primary cells The assay combines the YFP Venus halide sensor with a surrogate ion for chloride (iodide); upon stimulation of the chloride channel or transporter, iodide ions flow down the concentration gradient into the cells and quench YFP fluorescence upon binding; the amount of quench is directly proportional to the ion flux (chloride channel or transporter activity). Due to the bright Venus fluorescence, the Premo™ Halide Sensor signal window is excellent. We have demonstrated the utility and efficiency of the Premo™ Halide Sensor as a robust and true 'no wash' HTS chloride assay. Premo™ Halide Sensor is pre-packaged and ready for immediate use. It contains all components required for cellular delivery and expression. Screening can be conducted in complete medium and without any wash steps. To uncouple cell maintenance and preparation from screening, simply freeze transduced cells and bring out as little as 4 h before the screen. Both stable cell lines and human primary cells can be prepared frozen and 'assay-ready'.
TRUSTED_SUSTAINABILITY

Specifications

Color Yellow
Content And Storage The Premo Halide Sensor is a pharmacologically relevant sensor for functional studies of ligand- and voltage-gated chloride channels. It is based on a halide-sensitive form of Venus, the brightest available YFP molecule, affording an excellent signal window. Packaged in BacMam format, delivery is efficient in a range of cell types, including human primary cells. We have demonstrated the utility and efficiency of the Premo Halide Sensor as a robust and true 'no wash' HTS chloride assay.
Premo Halide Sensor should be stored at 4°C, protected from light and is supplied with an enhancer (store at 4°C). When stored as directed, the product is stable for 6 months.
Delivery Type Baculovirus (BacMam System)
Detection Method Fluorescence
For Use With (Application) Cell Viability and Proliferation
For Use With (Equipment) Fluorescence Microscope
Product Type Halide Sensor
Dye Type Protein-Based
Emission Visible
Form Liquid
Product Line Premo
Quantity 20 mL Kit
Shipping Condition Wet Ice
Target Ion Channels
Show More Show Less
How can I increase the transduction efficiency with the BacMam 2.0 reagents such as the the CellLight and Premo products?

Try varying particle-to-cell ratio (PPC), incubation volume, temperature and, cell density (if adherent cells are transduced). For adherent cells, we recommend a confluence of about 70%. Following the PPC, adjusting the volume is the next best parameter to change to optimize protein expression. If that doesn't work, you can also use the BacMam Enhancer Kit (Cat. No. B10107).

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.