Learn More
New Pig Tyvek™ IsoClean™ Cleanroom Boot Cover
SDP
Catalog No. 17987859 Shop All New Pig Corporation Products
Click to view available options
Size:
Small
Medium
Large
X-Large
Packaging Quantity:
100 Pk.
300 Pk.
These boot covers provide an excellent barrier to particles, microorganisms and non-hazardous light liquid splash in ISO Class 5-8 cleanrooms
  • Made from Tyvek™ material, which is a patented flashspun high-density polyethylene, these boot covers provide an inherent barrier to particles, microorganisms, and non-hazardous liquid splashes.
  • The fabric is specially processed to be exceptionally low-linting and to minimize particle shedding, which is a critical feature for maintaining the integrity of ISO Class 5-8 cleanrooms.
  • Tyvek™ is known for its durability. The material has a high abrasion resistance and is strong enough to withstand rigorous use without wearing away, ensuring reliable protection.
  • The boot covers are available in both clean-processed and sterile options.
  • The sterile versions are gamma-irradiated to a Sterility Assurance Level (SAL) of 10-6, making them suitable for GMP Class A/B (ISO Class 5) environments.
  • They are designed with features like tunnelled elastication at the shin and ankle, along with ankle ties, to provide a secure fit over footwear and prevent the boot cover from slipping or riding up.
  • The boot covers are often folded to facilitate aseptic donning, minimizing the risk of contamination during the gowning process.
Specifications
Size Small
Material Tyvek
Latex Free Yes
Color White
Disposable Single-use
Sterility Non-sterile
Closure Type Elastic Top with Ankle Ties
Thread Color White
Packaging Quantity 100 Pk.
Resistance Microorganisms, Dry Particulates, Splash
Product Type Controlled Environments Boot Covers
Cleanroom Class Class 100 (ISO 5)
Show More Show Less
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.