Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco™ Female Rep Sysm-Cervix Uteri
SCI_ED

Catalog No. S30391 Shop All Eisco Products
Click to view available options
Quantity:
1

Prepared microscope slide of a section of cervix uteri. Cervix uteri is a part of female reproductive system. Showing columnar epithelium in the canal of the cervix. Great choice for biology classrooms to explore structure function relationships as per NGSS standards.

  • pre-cleaned glass slide with ground edges
  • size 1x3 in. (25x77 mm)
  • professionally stained samples for best visualization
  • great choice for biology classrooms to explore structure-function relationships as per NGSS standards
  • showing columnar epithelium in the canal of the cervix

Specifications

Description Shows columnar epithelium in the canal of the cervix
Classification Female Reproductive System
Material Glass
Type Prepared Microscope Slide
For Use With (Application) Microscopy
Quantity 1
Specimen Type Cervix Uteri
Size 1 x 3 in.
Slide Type Prepared Slide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.