Learn More
Eisco™ Iris Stem Comination, Longitudinal Section & Cross Section
Shop All Eisco Products
Click to view available options
Quantity:
1 Ea.
Description
- Pre-cleaned glass slide with ground edges
- Size 1x3 in. (25x77 mm)
- Professionally stained samples for best visualization
- Great choice for biology classrooms to explore structure-function relationships as per NGSSs standards
- Showing characteristic monocot stem structures in two different points of view
Specifications
Specifications
Description | Cross and longitudinal sections of an Iris stem stained for better visualization of characteristic monocot structures |
Material | Glass |
Type | Prepared Microscope Slide |
For Use With (Application) | Microscopy |
Quantity | 1 Ea. |
Specimen Type | Iris Stem Combination |
Size | 1 x 3 in. |
Slide Type | Prepared Slide |