Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco™ Liver Section (Amphibian)
SCI_ED

Catalog No. S30596 Shop All Eisco Products
Click to view available options
Quantity:
1 Ea.

Catalog No. S30596

Supplier: Eisco™ BS18255

Only null left
Add to Cart
Add to Cart

Prepared microscope slide of an amphibian liver section. Stained for better visualization of characteristic structures such as blood vessels, sinusoids, and hepatic ducts. Great choice for biology classrooms to explore sturcture function relationships as per NGSS standards.

  • Pre-cleaned glass slide with ground edges
  • Size 1x3 in. (25x77 mm)
  • Professionally stained samples for best visualization
  • Great choice for biology classrooms to explore structure-function relationships as per NGSSs standards
  • Showing characteristic structures such as blood vessels, sinusoids, and hepatic ducts

Specifications

Description Amphibian Liver section stained for better visualization of blood vessels, sinusoids, and hepatic ducts
Material Glass
Type Prepared Microscope Slide
For Use With (Application) Microscopy
Quantity 1 Ea.
Species Amphibian
Specimen Type Liver
Size 1 x 3 in.
Slide Type Prepared Slide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.