Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Presenilin-2 Antibody (APS 21), Alexa Fluor™ 647, Novus Biologicals™
SDP

Catalog No. NB10074512Y Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10074512Y 0.2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10074512Y Supplier Novus Biologicals Supplier No. NB10074512AF647

Mouse Monoclonal Antibody

Presenilin-2 Monoclonal specifically detects Presenilin-2 in Human, Mouse, Rat, Primate samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen Presenilin-2
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone APS 21
Conjugate Alexa Fluor 647
Dilution Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias AD3LP, AD4AD3L, AD5, E5-1, EC 3.4.23, EC 3.4.23.-, presenilin 2 (Alzheimer disease 4), presenilin-2, PS-2, PS2CMD1V, PSNL2, STM2STM-2
Gene Symbols PSEN2
Host Species Mouse
Immunogen Synthetic peptide corresponding to residues C(31) Q E G R Q G P E D G E N T A(45) of human PS2.
Purification Method Protein G purified
Quantity 0.2 mL
Regulatory Status RUO
Research Discipline Alzheimers Research, Apoptosis, Neurodegeneration, Neuroscience, Phospho Specific, Stem Cell Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 5664
Test Specificity Presenilin 2 (APS 21)
Target Species Human, Mouse, Rat, Primate
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.