Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Preso/FRMPD4 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NB017346 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB017346 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB017346 Supplier Novus Biologicals Supplier No. NBP282035
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Preso/FRMPD4 Polyclonal specifically detects Preso/FRMPD4 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen Preso/FRMPD4
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1-2 μg/mL, ELISA, Immunohistochemistry 5 μg/mL, Immunocytochemistry/Immunofluorescence 20 μg/mL, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias FERM and PDZ domain containing 4, FERM and PDZ domain-containing protein 4, MGC142260, MGC142262, PDZ domain containing 10, PDZD10, PDZK10
Gene Symbols FRMPD4
Host Species Rabbit
Immunogen Antibody was raised against an 18 amino acid synthetic peptide from near the center of human FRMPD4. The immunogen is located within amino acids 620 - 670 of FRMPD4. Amino Acid Squence: LSGPETLKKAQESPRGAK
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9758
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.