Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PRG3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5110217
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5110217 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5110217 Supplier Invitrogen™ Supplier No. PA5110217
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LHLENDAPHL ESLETQADLG QDLDSSKEQE RDLALTEEVI QAEGEEVKAS ACQDNFEDEE AMESDPAALD KDFQCPREED IVEVQGSPRC KICRYLLVRT PKTFAEAQNV CSRCYGGNLV SIHDFNFNYR IQCCTSTVNQ AQVWIGGNLR GWFLWKRFCW TDGSHWNFAY WSPGQPGNGQ GSCVALCTKG GYWRRAQCDK QLPFVCSF.

Possesses similar cytotoxic and cytostimulatory activities to PRG2/MBP. In vitro, stimulates neutrophil superoxide production and IL8 release, and histamine and leukotriene C4 release from basophils. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen PRG3
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.69 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene PRG3
Gene Accession No. Q9JL95, Q9Y2Y8
Gene Alias eosinophil major basic protein 2; Eosinophil major basic protein homolog; major basic protein 2; MBP2; MBPH; Prepro-major basic protein homolog; prepro-MBPH; PRG3; proteoglycan 3; proteoglycan 3, pro eosinophil major basic protein 2; UNQ486/PRO1002
Gene Symbols PRG3
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 18-225 of human PRG3 (NP_006084.2).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10394, 295706, 53856
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.