Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PRICKLE1 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NB017082 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB017082 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB017082 Supplier Novus Biologicals Supplier No. NBP281771
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PRICKLE1 Polyclonal specifically detects PRICKLE1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen PRICKLE1
Applications Western Blot, ELISA, Immunofluorescence
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1 μg/mL, ELISA, Immunocytochemistry/Immunofluorescence 20 μg/mL
Formulation PBS
Gene Alias EPM1BMGC138902, FLJ31627, FLJ31937, prickle homolog 1 (Drosophila), prickle-like 1, prickle-like 1 (Drosophila), prickle-like protein 1, REST (RE-1 silencing transcription factor)/NRSF (neuron-restrictive silencerfactor)-interacting LIM domain protein, REST/NRSF-interacting LIM domain protein 1, RILPMGC138903
Gene Symbols PRICKLE1
Host Species Rabbit
Immunogen PRICKLE1 antibody was raised against a 19 amino acid synthetic peptide near the carboxy terminus of human PRICKLE1. The immunogen is located within the last 50 amino acids of PRICKLE1. Amino Acid Squence: LPTPQFGQRTTKSKKKKGH
Molecular Weight of Antigen 52, 90 kDa
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 144165
Test Specificity Multiple isoforms of PRICKLE1 are known to exist. PRICKLE1 antibody is predicted to not cross-react with other PRICKLE protein family members.
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.