Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PRKCBP1 Rabbit anti-Human, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB170459 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB170459 25 μg
NB170458 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB170459 Supplier Novus Biologicals Supplier No. NBP31864925UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PRKCBP1 Polyclonal antibody specifically detects PRKCBP1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), KnockDown

Specifications

Antigen PRKCBP1
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), KnockDown
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000, ELISA 1:100-1:500, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 0.25 to 2 μg/mL, Immunohistochemistry-Paraffin 1:20-1:50, Knockdown Validated
Formulation Whole antiserum
Gene Accession No. Q9ULU4
Gene Alias ESP-2, HED-2, zyxin, Zyxin-2
Host Species Rabbit
Immunogen Polyclonal antibody raised in rabbit against human PRKCBP1 (zinc finger, MYND-type containing 8), using a KLH-conjugated synthetic peptide containing a sequence from the C-terminal part of the protein.
Purification Method Whole Antiserum
Quantity 25 μg
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 23613
Target Species Human
Content And Storage Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles.
Form Antisera
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.