Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PRKCDBP Antibody, mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB342157 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB342157 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB342157 Supplier Novus Biologicals Supplier No. NBP281847MFV610
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PRKCDBP Polyclonal antibody specifically detects PRKCDBP in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen PRKCDBP
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate mFluor Violet 610 SE
Dilution Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias Cavin-3, hSRBC, MGC20400, protein kinase C, delta binding protein, sdr-related gene product that binds to c-kinase, Serum deprivation response factor-related gene product that binds to C-kinase, SRBCprotein kinase C delta-binding protein
Host Species Rabbit
Immunogen Rabbit polyclonal PRKCDBP antibody was raised against an 17 amino acid peptide near the center of human PRKCDBP. The immunogen is located within amino acids 150 - 200 of PRKCDBP. Amino Acid Squence: ESSDEEPVESRAQRLRRT
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Diabetes Research, Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 112464
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.