Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PRKCDBP Antibody, Novus Biologicals™
SDP

Catalog No. NBP144091 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP144091 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP144091 Supplier Novus Biologicals Supplier No. NBP144091
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 3 publications

PRKCDBP Polyclonal specifically detects PRKCDBP in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry.

Specifications

Antigen PRKCDBP
Applications Western Blot, ELISA, Immunohistochemistry
Classification Polyclonal
Concentration 1.02 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1ug/ml, ELISA 1:10000, Immunohistochemistry 5 ug/ml
Formulation 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 30% Glycerol with No Preservative
Gene Accession No. Q969G5
Gene Alias Cavin-3, hSRBC, MGC20400, protein kinase C, delta binding protein, sdr-related gene product that binds to c-kinase, Serum deprivation response factor-related gene product that binds to C-kinase, SRBCprotein kinase C delta-binding protein
Gene Symbols CAVIN3
Host Species Rabbit
Immunogen PRKCDBP Antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding to the C-terminal region of human PRKCDBP. (Uniprot: Q969G5)
Molecular Weight of Antigen 27.7 kDa
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Diabetes Research, Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 112464
Test Specificity A BLAST analysis was used to suggest cross-reactivity with rat, human, and mouse based on 100% sequence homology. Cross-reactivity with PRKCDBP from other sources has not been determined.
Reconstitution Reconstitute with 185 ml of water to desired concentration and for immediate usage.
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.