Learn More
PRKCSH Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A304745AT

Description
The recommended shelf life for this product is 1 year from date of receipt.
This gene encodes the beta-subunit of glucosidase II, an N-linked glycan-processing enzyme in the endoplasmic reticulum (ER). This protein is an acidic phospho-protein known to be a substrate for protein kinase C. Mutations in this gene have been associated with the autosomal domit polycystic liver disease (PCLD). Alternatively spliced transcript variants encoding distinct isoforms have been observed.Specifications
PRKCSH | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
O08795, P14314 | |
PRKCSH | |
Between 1 and 50 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
PRKCSH | |
AGE-R2, G19P1, PCLD, PKCSH, PLD1, AGE-binding receptor 2, glucosidase 2 subunit beta, glucosidase II subunit beta, glucosidase II, beta subunit, hepatocystin, protein kinase C substrate 60.1 kDa protein heavy chain, protein kinase C substrate, 80 Kda protein | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
19089, 5589 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.