Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PRKCSH Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
$151.31
Specifications
Antigen | PRKCSH |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Liquid |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-156-8557
![]() |
Bethyl Laboratories, Inc
A304745AT |
20 μL |
Each of 1 for $151.31
|
|
|||||
Description
The recommended shelf life for this product is 1 year from date of receipt.
This gene encodes the beta-subunit of glucosidase II, an N-linked glycan-processing enzyme in the endoplasmic reticulum (ER). This protein is an acidic phospho-protein known to be a substrate for protein kinase C. Mutations in this gene have been associated with the autosomal domit polycystic liver disease (PCLD). Alternatively spliced transcript variants encoding distinct isoforms have been observed.Specifications
PRKCSH | |
Polyclonal | |
Liquid | |
RUO | |
TBS with 0.1% BSA and 0.09% sodium azide | |
AGE-R2, G19P1, PCLD, PKCSH, PLD1, AGE-binding receptor 2, glucosidase 2 subunit beta, glucosidase II subunit beta, glucosidase II, beta subunit, hepatocystin, protein kinase C substrate 60.1 kDa protein heavy chain, protein kinase C substrate, 80 Kda protein | |
Between 1 and 50 | |
Primary | |
4° C | |
PRKCSH |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
O08795, P14314 | |
19089, 5589 | |
IgG | |
Antigen affinity chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title