Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PRKCSH Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB167730 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB167730 20 μg
NB167729 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB167730 Supplier Novus Biologicals Supplier No. NBP31631920UL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PRKCSH Polyclonal antibody specifically detects PRKCSH in Human, Mouse, Rat samples. It is validated for Western Blot

Specifications

Antigen PRKCSH
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS, 50% glycerol, pH7.3
Gene Alias AGE-R2,80K-H protein, G19P1AGE-binding receptor 2, glucosidase 2 subunit beta, Glucosidase II subunit beta, glucosidase II, beta subunit, hepatocystin, PCLD, PKCSH, PLD1, Protein kinase C substrate 60.1 kDa protein heavy chain, protein kinase C substrate 80K-H, protein kinase C substrate, 80 Kda protein
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 130-275 of human PRKCSH (P14314).
Purification Method Affinity purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 5589
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.