Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PRKCSH Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$148.20 - $378.00
Specifications
Antigen | PRKCSH |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Description
PRKCSH Polyclonal antibody specifically detects PRKCSH in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
PRKCSH | |
Polyclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
AGE-R2,80K-H protein, G19P1AGE-binding receptor 2, glucosidase 2 subunit beta, Glucosidase II subunit beta, glucosidase II, beta subunit, hepatocystin, PCLD, PKCSH, PLD1, Protein kinase C substrate 60.1 kDa protein heavy chain, protein kinase C substrate 80K-H, protein kinase C substrate, 80 Kda protein | |
Recombinant fusion protein containing a sequence corresponding to amino acids 130-275 of human PRKCSH (P14314). | |
Primary | |
Store at 4°C. |
Western Blot | |
Unconjugated | |
Rabbit | |
Signal Transduction | |
PBS, 50% glycerol, pH7.3 | |
5589 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title